Learn More
Abnova™ Human MRPL44 Full-length ORF (NP_075066.1, 1 a.a. - 332 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00065080P01S
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. [provided by RefSeq]
Sequence: MASGLVRLLQQGHRCLLAPVAPKLVPPVRGVKKGFRAAFRFQKELERQRLLRCPPPPVRRSEKPNWDYHAEIQAFGHRLQENFSLDLLKTAFVNSCYIKSEEAKRQQLGIEKEAVLLNLKSNQELSEQGTSFSQTCLTQFLEDEYPDMPTEGIKNLVDFLTGEEVVCHVARNLAVEQLTLSEEFPVPPAVLQQTFFAVIGALLQSSGPERTALFIRDFLITQMTGKELFEMWKIINPMGLLVEELKKRNVSAPESRLTRQSGGTTALPLYFVGLYCDKKLIAEGPGETVLVAEEEAARVALRKLYGFTENRRPWNYSKPKETLRAEKSITASSpecifications
NP_075066.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12701/FLJ13990/FLJ37688/L44MT/MRP-L44 | |
MRPL44 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
65080 | |
MRPL44 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASGLVRLLQQGHRCLLAPVAPKLVPPVRGVKKGFRAAFRFQKELERQRLLRCPPPPVRRSEKPNWDYHAEIQAFGHRLQENFSLDLLKTAFVNSCYIKSEEAKRQQLGIEKEAVLLNLKSNQELSEQGTSFSQTCLTQFLEDEYPDMPTEGIKNLVDFLTGEEVVCHVARNLAVEQLTLSEEFPVPPAVLQQTFFAVIGALLQSSGPERTALFIRDFLITQMTGKELFEMWKIINPMGLLVEELKKRNVSAPESRLTRQSGGTTALPLYFVGLYCDKKLIAEGPGETVLVAEEEAARVALRKLYGFTENRRPWNYSKPKETLRAEKSITAS | |
RUO | |
MRPL44 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.