Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MRPS9 Full-length ORF (NP_872578.1, 1 a.a. - 396 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00064965P01L

 View more versions of this product

Catalog No. 89-952-993


Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. [provided by RefSeq]

Sequence: MAAPCVSYGGAVSYRLLLWGRGSLARKQGLWKTAAPELQTNVRSQILRLRHTAFVIPKKNVPTSKRETYTEDFIKKQIEEFNIGKRHLANMMGEDPETFTQEDIDRAIAYLFPSGLFEKRARPVMKHPEQIFPRQRAIQWGEDGRPFHYLFYTGKQSYYSLMHDVYGMLLNLEKHQSHLQAKSLLPEKTVTRDVIGSRWLIKEELEEMLVEKLSDLDYMQFIRLLEKLLTSQCGAAEEEFVQRFRRSVTLESKKQLIEPVQYDEQGMAFSKSEGKRKTAKAEAIVYKHGSGRIKVNGIDYQLYFPITQDREQLMFPFHFVDRLGKHDVTCTVSGGGRSAQAGAIRLAMAKALCSFVTEDEVEWMRQAGLLTTDPRVRERKKPGQEGARRKFTWKKR
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
72.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MRP-S9/RPMS9/S9mt
MRPS9
Yes
NP_872578.1
Liquid
64965
MRPS9 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAPCVSYGGAVSYRLLLWGRGSLARKQGLWKTAAPELQTNVRSQILRLRHTAFVIPKKNVPTSKRETYTEDFIKKQIEEFNIGKRHLANMMGEDPETFTQEDIDRAIAYLFPSGLFEKRARPVMKHPEQIFPRQRAIQWGEDGRPFHYLFYTGKQSYYSLMHDVYGMLLNLEKHQSHLQAKSLLPEKTVTRDVIGSRWLIKEELEEMLVEKLSDLDYMQFIRLLEKLLTSQCGAAEEEFVQRFRRSVTLESKKQLIEPVQYDEQGMAFSKSEGKRKTAKAEAIVYKHGSGRIKVNGIDYQLYFPITQDREQLMFPFHFVDRLGKHDVTCTVSGGGRSAQAGAIRLAMAKALCSFVTEDEVEWMRQAGLLTTDPRVRERKKPGQEGARRKFTWKKR
RUO
MRPS9
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit