Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MRPS9 Full-length ORF (NP_872578.1, 1 a.a. - 396 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name MRPS9 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 72.2kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-952-992
View Documents
Abnova™
H00064965P01S
10 ug
Each for $460.50
Add to cart
 
89-952-993
View Documents
Abnova™
H00064965P01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. [provided by RefSeq]

Sequence: MAAPCVSYGGAVSYRLLLWGRGSLARKQGLWKTAAPELQTNVRSQILRLRHTAFVIPKKNVPTSKRETYTEDFIKKQIEEFNIGKRHLANMMGEDPETFTQEDIDRAIAYLFPSGLFEKRARPVMKHPEQIFPRQRAIQWGEDGRPFHYLFYTGKQSYYSLMHDVYGMLLNLEKHQSHLQAKSLLPEKTVTRDVIGSRWLIKEELEEMLVEKLSDLDYMQFIRLLEKLLTSQCGAAEEEFVQRFRRSVTLESKKQLIEPVQYDEQGMAFSKSEGKRKTAKAEAIVYKHGSGRIKVNGIDYQLYFPITQDREQLMFPFHFVDRLGKHDVTCTVSGGGRSAQAGAIRLAMAKALCSFVTEDEVEWMRQAGLLTTDPRVRERKKPGQEGARRKFTWKKR
Specifications

Specifications

MRPS9 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
72.2kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit