Learn More
Abnova™ Human MSC Partial ORF (NP_005089.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00009242Q02S
Description
The protein encoded by this gene is a transcriptional repressor capable of binding an E-box element either as a homodimer or as a heterodimer with E2A in vitro. The encoded protein also forms heterodimers with E2A proteins in vivo. This protein is capable of inhibiting the transactivation capability of E47, an E2A protein, in mammalian cells. This gene is a downstream target of the B-cell receptor signal transduction pathway. [provided by RefSeq]
Sequence: MSTGSVSDPEEMELRGLQREYPVPASKRPPLRGVERSYASPSDNSSAEEEDPDGEEERCALGTAGSAEGCKRKRPRVAGGGGAGGSAGGGGKKPLPAKGSSpecifications
NP_005089.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSTGSVSDPEEMELRGLQREYPVPASKRPPLRGVERSYASPSDNSSAEEEDPDGEEERCALGTAGSAEGCKRKRPRVAGGGGAGGSAGGGGKKPLPAKGS | |
RUO | |
MSC | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9242 | |
MSC (Human) Recombinant Protein (Q02) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ABF-1/ABF1/MYOR/bHLHa22 | |
MSC | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.