Learn More
Abnova™ Human MSL3L1 Partial ORF (NP_523353.1, 1 a.a. - 79 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00010943Q01L
Description
This gene encodes a nuclear protein and has similarity to drosophila male-specific lethal-3 gene. The drosophila protein plays a critical role in a dosage-compensation pathway, which equalizes X-linked gene expression in males and females. Thus this encoded protein is thought to play a similar function in chromatin remodeling and transcriptional regulation. This gene has been found to undergo X inactivation. There are four alternatively spliced transcript variants of this gene encoding different isoforms. [provided by RefSeq]
Sequence: MSASEGMKFKFHSGEKVLCFEPDPTKARVLYDAKIVVVIVGKDEKGRKIPEYLIHFNGWNRSWDRWAAEDHVLRDTDENSpecifications
NP_523353.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
34.43kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MSASEGMKFKFHSGEKVLCFEPDPTKARVLYDAKIVVVIVGKDEKGRKIPEYLIHFNGWNRSWDRWAAEDHVLRDTDEN | |
RUO | |
MSL3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
10943 | |
MSL3L1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586J1822/MSL3L1 | |
MSL3 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.