Learn More
Abnova™ Human MTMR9 Partial ORF (NP_056273, 450 a.a. - 549 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00066036Q01S
Description
This gene encodes a myotubularin-related protein that is atypical to most other members of the myotubularin-related protein family because it has no dual-specificity phosphatase domain. The encoded protein contains a double-helical motif similar to the SET interaction domain, which is thought to have a role in the control of cell proliferation. In mouse, a protein similar to the encoded protein binds with MTMR7, and together they dephosphorylate phosphatidylinositol 3-phosphate and inositol 1,3-bisphosphate. [provided by RefSeq]
Sequence: LQQKTMSLWSWVNQPSELSKFTNPLFEANNLVIWPSVAPQSLPLWEGIFLRWNRSSKYLDEAYEEMVNIIEYNKELQAKVNILRRQLAELETEDGMQESPSpecifications
NP_056273 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LQQKTMSLWSWVNQPSELSKFTNPLFEANNLVIWPSVAPQSLPLWEGIFLRWNRSSKYLDEAYEEMVNIIEYNKELQAKVNILRRQLAELETEDGMQESP | |
RUO | |
MTMR9 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
66036 | |
MTMR9 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
C8orf9/DKFZp434K171/LIP-STYX/MGC126672/MTMR8 | |
MTMR9 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.