Learn More
Abnova™ Human MTRF1 Full-length ORF (ENSP00000239852, 1 a.a. - 151 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | ENSP00000239852 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 9617 |
Molecular Weight (g/mol) | 44.6kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-953-212
|
Abnova™
H00009617P01L |
25 ug |
Each for $769.00
|
|
|||||
89-953-211
|
Abnova™
H00009617P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
The protein encoded by this gene was determined by in silico methods to be a mitochondrial protein with similarity to the peptide chain release factors (RFs) discovered in bacteria and yeast. The peptide chain release factors direct the termination of translation in response to the peptide chain termination codons. Initially thought to have a role in the termination of mitochondria protein synthesis, a recent publication found no mitochondrial translation release functionality. Multiple alternatively spliced transcript variants have been suggested by mRNA and EST data; however, their full-length natures are not clear. [provided by RefSeq]
Sequence: MNRHLCVWLFRHPSLNGYLQCHIQLHSHQFRQIHLDTRLQVFRQNRNCILHLLSKNWSRRYCHQDTKMLWKHKALQKYMENLSKEYQTLEQCLQHIPVNEENRRSLNRRHAELAPLAAIYQEIQETEQAIEELESMCKKTESCSVAQAGMQSpecifications
ENSP00000239852 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
44.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNRHLCVWLFRHPSLNGYLQCHIQLHSHQFRQIHLDTRLQVFRQNRNCILHLLSKNWSRRYCHQDTKMLWKHKALQKYMENLSKEYQTLEQCLQHIPVNEENRRSLNRRHAELAPLAAIYQEIQETEQAIEELESMCKKTESCSVAQAGMQ | |
RUO | |
MTRF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9617 | |
MTRF1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC47721/MRF1/MTTRF1/RF1 | |
MTRF1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.