Learn More
Novus Biologicals™ Human MUC21 ELISA Kit (Colorimetric)
Human MUC21 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276677
Catalog No. NBP276677
Add to cart
Description
Description
The Human MUC21 ELISA Kit (Colorimetric) measures human MUC21 in serum, plasma and other biological fluids.
Specifications
Specifications
0.156 to 10 ng/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
MUC21
Biological Fluids, Plasma, Serum
394263
0.094 ng/mL
Human
1 Kit