Learn More
Novus Biologicals™ Human MUC7 ELISA Kit (Colorimetric)
Human MUC7 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276707
Catalog No. NBP276707
Add to cart
Description
Description
The Human MUC7 ELISA Kit (Colorimetric) measures human MUC7 in serum, plasma and other biological fluids.
Specifications
Specifications
0.313 to 20 ng/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
MUC7
Biological Fluids, Plasma, Serum
4589
0.188 ng/mL
Human
1 Kit