Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human MUCDHL Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3826SP
This item is not returnable.
View return policy
Description
MUCDHL Polyclonal specifically detects MUCDHL in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MUCDHL | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cadherin-related family member 5, CDHR5, FLJ20219, MUCDHLmucin-like protocadherin, mucin and cadherin-like, Mucin and cadherin-like protein, MUPCDH, MU-PCDH, mu-protocadherin | |
Goat | |
Affinity Purified | |
RUO | |
53841 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Q9HBB8 | |
CDHR5 | |
E. coli-derived recombinant human MUCDHL Gln26-Ala434 Accession # Q9HBB8 | |
25 μg | |
Primary | |
Detects human MUCDHL in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 10% cross-reactivity with recombinant mouse MUCDHL is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction