Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human MUTED Partial ORF (NP_958437.1, 88 a.a. - 187 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Supplier:  Abnova™ H00063915Q01L

 View more versions of this product

Catalog No. 89-953-268


Add to Cart

Description

Description

This gene encodes a component of BLOC-1 (biogenesis of lysosome-related organelles complex 1). Components of this complex are involved in the biogenesis of organelles such as melanosomes and platelet-dense granules. A mouse model for Hermansky-Pudlak Syndrome is mutated in the murine version of this gene. Some transcripts of the downstream gene TXNDC5 overlap this gene, but they do not contain an open reading frame for this gene. [provided by RefSeq]

Sequence: EHTLPKCRDTMRDSLSQVLQRLQAANDSVCRLQQREQERKKIHSDHLVASEKQHMLQWDNFMKEQPNKRAEVDEEHRKAMERLKEQYAEMEKDLAKFSTF
Specifications

Specifications

NP_958437.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EHTLPKCRDTMRDSLSQVLQRLQAANDSVCRLQQREQERKKIHSDHLVASEKQHMLQWDNFMKEQPNKRAEVDEEHRKAMERLKEQYAEMEKDLAKFSTF
RUO
MUTED
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
63915
MUTED (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686E2287/MU
MUTED
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit