Learn More
Abnova™ Human MYO15A Partial ORF (NP_057323.2, 3432 a.a. - 3530 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00051168Q01S
Description
This gene encodes an unconventional myosin. This protein differs from other myosins in that it has a long N-terminal extension preceding the conserved motor domain. Studies in mice suggest that this protein is necessary for actin organization in the hair cells of the cochlea. Mutations in this gene have been associated with profound, congenital, neurosensory, nonsyndromal deafness. This gene is located within the Smith-Magenis syndrome region on chromosome 17. Read-through transcripts containing an upstream gene and this gene have been identified, but they are not thought to encode a fusion protein. Several alternatively spliced transcript variants have been described, but their full length sequences have not been determined. [provided by RefSeq]
Sequence: PCILAINHNGLNFLSTETHELMVKFPLKEIQSTRTQRPTANSSYPYVEIALGDVAAQRTLQLQLEQGLELCRVVAVHVENLLSAHEKRLTLPPSEITLLSpecifications
NP_057323.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PCILAINHNGLNFLSTETHELMVKFPLKEIQSTRTQRPTANSSYPYVEIALGDVAAQRTLQLQLEQGLELCRVVAVHVENLLSAHEKRLTLPPSEITLL | |
RUO | |
MYO15A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
51168 | |
MYO15A (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DFNB3/DKFZp686N18198/FLJ17274/FLJ31311/MYO15 | |
MYO15A | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.