Learn More
Novus Biologicals™ Human N-MID Osteocalcin ELISA Kit (Colorimetric)
This kit recognizes Human N-MID-OT in samples.
Supplier: Novus Biologicals™ NBP279696
Description
No Significant cross-reactivity or interference between Human N-MID-OT and analogues was observed.Specifications
0.63 to 40 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
Human | |
Storage of components varies. See protocol for specific instructions. |
0.38 ng/mL | |
Other Biological Fluids, Plasma, Serum | |
632 | |
1 Kit | |
N-MID Osteocalcin |