Learn More
Abnova™ Human NAGK Full-length ORF (AAH01029, 1 a.a. - 344 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH01029 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55577 |
Molecular Weight (g/mol) | 63.58kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-953-554
|
Abnova™
H00055577P01S |
10 ug |
Each for $499.00
|
|
|||||
89-953-555
|
Abnova™
H00055577P01L |
25 ug |
Each for $769.00
|
|
|||||
Description
N-acetylglucosamine kinase (NAGK; EC 2.7.1.59) converts endogenous N-acetylglucosamine (GlcNAc), a major component of complex carbohydrates, from lysosomal degradation or nutritional sources into GlcNAc 6-phosphate. NAGK belongs to the group of N-acetylhexosamine kinases and is a prominent salvage enzyme of amino sugar metabolism in mammals.[supplied by OMIM]
Sequence: MAAIYGGVEGGGTRSEVLLVSEDGKILAEADGLSTNHWLIGTDKCVERINEMVNRAKRKAGVDPLVPLRSLGLSLSGGDQEDAGRILIEELRDRFPYLSESYLITTDAAGSIATATPDGGVVLISGTGSNCRLINPDGSESGCGGWGHMMGDEGSAYWIAHQAVKIVFDSIDNLEAAPHDIGYVKQAMFHYFQVPDRLGILTHLYRDFDKCRFAGFCRKIAEGAQQGDPLSRYIFRKAGEMLGRHIVAVLPEIDPVLFQGKIGLPILCVGSVWKSWELLKEGFLLALTQGREIQAQNFFSSFTLMKLRHSSALGGASLGARHIGHLLPMDYSANAIAFYSYTFSSpecifications
AAH01029 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
63.58kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAIYGGVEGGGTRSEVLLVSEDGKILAEADGLSTNHWLIGTDKCVERINEMVNRAKRKAGVDPLVPLRSLGLSLSGGDQEDAGRILIEELRDRFPYLSESYLITTDAAGSIATATPDGGVVLISGTGSNCRLINPDGSESGCGGWGHMMGDEGSAYWIAHQAVKIVFDSIDNLEAAPHDIGYVKQAMFHYFQVPDRLGILTHLYRDFDKCRFAGFCRKIAEGAQQGDPLSRYIFRKAGEMLGRHIVAVLPEIDPVLFQGKIGLPILCVGSVWKSWELLKEGFLLALTQGREIQAQNFFSSFTLMKLRHSSALGGASLGARHIGHLLPMDYSANAIAFYSYTFS | |
RUO | |
NAGK | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55577 | |
NAGK (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GNK/HSA242910 | |
NAGK | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.