Learn More
Abnova™ Human NANS Full-length ORF (NP_061819.2, 1 a.a. - 359 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00054187P01S
Description
This gene encodes an enzyme that functions in the biosynthetic pathways of sialic acids. In vitro, the encoded protein uses N-acetylmannosamine 6-phosphate and mannose 6-phosphate as substrates to generate phosphorylated forms of N-acetylneuraminic acid (Neu5Ac) and 2-keto-3-deoxy-D-glycero-D-galacto-nononic acid (KDN), respectively; however, it exhibits much higher activity toward the Neu5Ac phosphate product. In insect cells, expression of this gene results in Neu5Ac and KDN production. This gene is related to the E. coli sialic acid synthase gene neuB, and it can partially restore sialic acid synthase activity in an E. coli neuB-negative mutant. [provided by RefSeq]
Sequence: MPLELELCPGRWVGGQHPCFIIAEIGQNHQGDLDVAKRMIRMAKECGADCAKFQKSELEFKFNRKALERPYTSKHSWGKTYGEHKRHLEFSHDQYRELQRYAEEVGIFFTASGMDEMAVEFLHELNVPFFKVGSGDTNNFPYLEKTAKKGRPMVISSGMQSMDTMKQVYQIVKPLNPNFCFLQCTSAYPLQPEDVNLRVISEYQKLFPDIPIGYSGHETGIAISVAAVALGAKVLERHITLDKTWKGSDHSASLEPGELAELVRSVRLVERALGSPTKQLLPCEMACNEKLGKSVVAKVKIPEGTILTMDMLTVKVGEPKGYPPEDIFNLVGKKVLVTVEEDDTIMEELVDNHGKKIKSSpecifications
NP_061819.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
66.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SAS | |
NANS | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54187 | |
NANS (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MPLELELCPGRWVGGQHPCFIIAEIGQNHQGDLDVAKRMIRMAKECGADCAKFQKSELEFKFNRKALERPYTSKHSWGKTYGEHKRHLEFSHDQYRELQRYAEEVGIFFTASGMDEMAVEFLHELNVPFFKVGSGDTNNFPYLEKTAKKGRPMVISSGMQSMDTMKQVYQIVKPLNPNFCFLQCTSAYPLQPEDVNLRVISEYQKLFPDIPIGYSGHETGIAISVAAVALGAKVLERHITLDKTWKGSDHSASLEPGELAELVRSVRLVERALGSPTKQLLPCEMACNEKLGKSVVAKVKIPEGTILTMDMLTVKVGEPKGYPPEDIFNLVGKKVLVTVEEDDTIMEELVDNHGKKIKS | |
RUO | |
NANS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.