Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NCKIPSD Full-length ORF (NP_909119.1, 1 a.a. - 715 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name NCKIPSD (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 104.6kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-953-725
View Documents
Abnova™
H00051517P01L
25 ug
Each for $709.50
Add to cart
 
89-953-724
View Documents
Abnova™
H00051517P01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

The protein encoded by this gene is localized exclusively in the cell nucleus. It plays a role in signal transduction, and may function in the maintenance of sarcomeres and in the assembly of myofibrils into sarcomeres. It also plays an important role in stress fiber formation. The gene is involved in therapy-related leukemia by a chromosomal translocation t(3;11)(p21;q23) that involves this gene and the myeloid/lymphoid leukemia gene. Alternative splicing occurs in this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]

Sequence: MYRALYAFRSAEPNALAFAAGETFLVLERSSAHWWLAARARSGETGYVPPAYLRRLQGLEQDVLQAIDRAIEAVHNTAMRDGGKYSLEQRGVLQKLIHHRKETLSRRGPSASSVAVMTSSTSDHHLDAAAARQPNGVCRAGFERQHSLPSSEHLGADGGLYQIPPQPRRAAPTTPPPPVKRRDREALMASGSGGHNTMPSGGNSVSSGSSVSSTSLDTLYTSSSPSEPGSSCSPTPPPVPRRGTHTTVSQVQPPPSKASAPEPPAEEEVATGTTSASDDLEALGTLSLGTTEEKAAAEAAVPRTIGAELMELVRRNTGLSHELCRVAIGIIVGHIQASVPASSPVMEQVLLSLVEGKDLSMALPSGQVCHDQQRLEVIFADLARRKDDAQQRSWALYEDEGVIRCYLEELLHILTDADPEVCKKMCKRNEFESVLALVAYYQMEHRASLRLLLLKCFGAMCSLDAAIISTLVSSVLPVELARDMQTDTQDHQKLCYSALILAMVFSMGEAVPYAHYEHLGTPFAQFLLNIVEDGLPLDTTEQLPDLCVNLLLALNLHLPAADQNVIMAALSKHANVKIFSEKLLLLLNRGDDPVRIFKHEPQPPHSVLKFLQDVFGSPATAAIFYHTDMMALIDITVRHIADLSPGDKLRMEYLSLMHAIVRTTPYLQHRHRLPDLQAILRRILNEEETSPQCQMDRMIVREMCKEFLVLGEAPS
Specifications

Specifications

NCKIPSD (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
104.6kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit