Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NCKIPSD Partial ORF (NP_909119.1, 1 a.a. - 99 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051517Q01L

 View more versions of this product

Catalog No. 89-953-723


Add to cart

Description

Description

The protein encoded by this gene is localized exclusively in the cell nucleus. It plays a role in signal transduction, and may function in the maintenance of sarcomeres and in the assembly of myofibrils into sarcomeres. It also plays an important role in stress fiber formation. The gene is involved in therapy-related leukemia by a chromosomal translocation t(3;11)(p21;q23) that involves this gene and the myeloid/lymphoid leukemia gene. Alternative splicing occurs in this locus and two transcript variants encoding distinct isoforms have been identified. [provided by RefSeq]

Sequence: MYRALYAFRSAEPNALAFAAGETFLVLERSSAHWWLAARARSGETGYVPPAYLRRLQGLEQDVLQAIDRAIEAVHNTAMRDGGKYSLEQRGVLQKLIHH
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MYRALYAFRSAEPNALAFAAGETFLVLERSSAHWWLAARARSGETGYVPPAYLRRLQGLEQDVLQAIDRAIEAVHNTAMRDGGKYSLEQRGVLQKLIHH
RUO
NCKIPSD
Wheat Germ (in vitro)
GST
NP_909119.1
Liquid
51517
NCKIPSD (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
AF3P21/DIP/DIP1/MGC23891/ORF1/SPIN90/WASLBP/WISH
NCKIPSD
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit