Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human NDOR1 Full-length ORF (XP_941628.1, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00027158P02S
Description
This gene encodes an NADPH-dependent diflavin reductase that contains both flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) binding domains. The encoded protein is an enzyme that catalyzes the transfers electrons from NADPH through FAD and FMN cofactors to potential redox partners. Alternative splicing results in multiple transcript variants. [provided by RefSeq]
Sequence: MEGAWALPTWKEEGREQAAGQGEEEECPICTEPYGPRERRLALLNCSHGLCVGCLHRLLGSASSADLGRVRCPLCRQKTPVLEWEICRLQEELLQADGPSRQPRREAPASYHRNPGPWGSLEHRYQLRFLAGPVGGRGCLPFLPCPPCLGARLWTLRERGPCARRLALLSLLALELLGLLLVFTPLLLLGLLFVLLDRSGRSpecifications
XP_941628.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC138148/NR1/bA350O14.9 | |
NDOR1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
27158 | |
NDOR1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEGAWALPTWKEEGREQAAGQGEEEECPICTEPYGPRERRLALLNCSHGLCVGCLHRLLGSASSADLGRVRCPLCRQKTPVLEWEICRLQEELLQADGPSRQPRREAPASYHRNPGPWGSLEHRYQLRFLAGPVGGRGCLPFLPCPPCLGARLWTLRERGPCARRLALLSLLALELLGLLLVFTPLLLLGLLFVLLDRSGR | |
RUO | |
NDOR1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction