Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NDOR1 Full-length ORF (XP_941628.1, 1 a.a. - 201 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00027158P02L

 View more versions of this product

Catalog No. 89-953-774



Description

Description

This gene encodes an NADPH-dependent diflavin reductase that contains both flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) binding domains. The encoded protein is an enzyme that catalyzes the transfers electrons from NADPH through FAD and FMN cofactors to potential redox partners. Alternative splicing results in multiple transcript variants. [provided by RefSeq]

Sequence: MEGAWALPTWKEEGREQAAGQGEEEECPICTEPYGPRERRLALLNCSHGLCVGCLHRLLGSASSADLGRVRCPLCRQKTPVLEWEICRLQEELLQADGPSRQPRREAPASYHRNPGPWGSLEHRYQLRFLAGPVGGRGCLPFLPCPPCLGARLWTLRERGPCARRLALLSLLALELLGLLLVFTPLLLLGLLFVLLDRSGR
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.8kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC138148/NR1/bA350O14.9
NDOR1
Yes
XP_941628.1
Liquid
27158
NDOR1 (Human) Recombinant Protein (P02)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEGAWALPTWKEEGREQAAGQGEEEECPICTEPYGPRERRLALLNCSHGLCVGCLHRLLGSASSADLGRVRCPLCRQKTPVLEWEICRLQEELLQADGPSRQPRREAPASYHRNPGPWGSLEHRYQLRFLAGPVGGRGCLPFLPCPPCLGARLWTLRERGPCARRLALLSLLALELLGLLLVFTPLLLLGLLFVLLDRSGR
RUO
NDOR1
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit