Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NEK2 (NM_002497) Full-length Recombinant Protein with GST-tag expressed in Sf9 cells

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P3884

Catalog No. 89-953-953

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Insect cell (Sf9) expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
76kDa
Insect cell (Sf9) expression system
SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HsPK21|NEK2A|NLK1
NEK2
Insect
GST
NM_002497
Liquid
4751
NEK2 (Human) Recombinant Protein
>90% by densitometry
5 ug
RUO
NEK2
The specific activity was determined to be 269nmol/min/mg as per Activity Assay Protocol.
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit