Learn More
Abnova™ Human NEK6 (Q9HC98) Full-length Recombinant Protein with GST-tag expressed in Sf9 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4200
Catalog No. 89-953-969
Add to cart
Description
Description
The Aspergillus nidulans 'never in mitosis A' (NIMA) gene encodes a serine/threonine kinase that controls initiation of mitosis. NIMA-related kinases (NEKs) are a group of protein kinases that are homologous to NIMA. Evidence suggests that NEKs perform functions similar to those of NIMA.[supplied by OMIM]
Specifications
Specifications
Q9HC98
Liquid
10783
NEK6 (Human) Recombinant Protein
1 ug/lane SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SID6-1512
NEK6
Insect
GST
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
63kDa
≥90% by SDS-PAGE
5 ug
RUO
NEK6
153nmol phosphate incorporated into MBP per minute per mg protein at 30°C for 15 minutes using a final concentration of 50μM ATP (0.83 uCi/assay).
Yes