Learn More
Raybiotech IncSupplier Diversity Partner Human/Mouse/Rat Neurokinin-B EIA Kit. DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner EIANEB1
Optimized for serum and cell culture medium. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8824