Learn More
Novus Biologicals™ Human Neuropeptide Y ELISA Kit (Colorimetric)
SDP
Human Neuropeptide Y ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Supplier:  Novus Biologicals™ NBP276680
Catalog No. NBP276680
Add to Cart
Description
Description
The Human Neuropeptide Y ELISA Kit (Colorimetric) measures human Neuropeptide Y in serum, plasma and other biological fluids.
Specifications
Specifications
31.25 to 2000 pg/mL
ELISA Kit (Colorimetric)
4852
Human
Neuroscience, Neurotransmission
18.75 pg/mL
Biological Fluids, Plasma, Serum
NPY
1 Kit
Storage of components varies. See protocol for specific instructions.