Learn More
BioworldSupplier Diversity Partner HUMAN NEUROTROPHIN-3 (RECOM DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 220604301
HUMAN NEUROTROPHIN-3 (RECOMBINANT): Recombinant Human Neurotrophin-3 (NT-3) Source: E.Coli Background: NT-3 is a neurotrophic factor involved in regulating the survival of visceral and proprioceptive sensory neurons. It is closely ho...
Catalog No. 50-253-527