Learn More
Gibco™ Human Neurturin Recombinant Protein, PeproTech®
SDP
E. coli Recombinant Protein
Supplier:  Gibco™ 4501120UG
Encompass
 View more versions of this product
Catalog No. 450-11-20UG
Add to Cart
This item is not returnable. View return policy
Description
Description
Glial cell line-derived neurotrophic factor (GDNF) plays key roles in the control of vertebrate neuron survival and differentiation. A novel neurotrophic factor was recently cloned from human and mouse and designated neurturin. Physiological responses to neurturin (NTN) require the presence of receptor tyrosine kinase RET and a novel glycosylphosphatidylinositol linked receptor NTNRalpha. The cDNAs encoding NTNRalpha from human, rat, chicken, and mouse have been cloned recently and termed GDNFR-b, Ret ligand 2 (RETL2) or TGF-beta-related neurotrophic factor receptor 2 (TrnR2) and nominated as GFR-a-2 recently. NTN binds to and forms a complex with GFR-a-2 and the Ret PTK and activates the RET receptor tyrosine kinase pathway. Both NTN and GDNF can activate the MAP kinase and phosphatidylinositol 3-kinase pathways and play a critical role in the development of many neuronal populations. Neurturin and GDNF define a new family of neurotrophic factors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Q99748
protein with no preservative
23.6
20 μg
-20°C
<1 EU/μg
NRTN
Unconjugated
ARLGARPCGL RELEVRVSEL GLGYASDETV LFRYCAGACE AAARVYDLGL RRLRQRRRLR RERVRAQPCC RPTAYEDEVS FLDAHSRYHT VHELSARECA CV
≥ 98% by SDS-PAGE gel and HPLC analyses.
ELISA, Functional Assay, Western Blot
4902
Human Neurturin
E. coli
RUO
neurturin; NRTN; NTN; prepro-neurturin
Human
Recombinant
Lyophilized
Product Suggestions
Product Suggestions