Learn More
Novus Biologicals™ Human Neutrophil Elastase/ELA2 ELISA Kit
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture. 
Manufacturer:  Novus Biologicals™ NBP191266
Catalog No. NBP191266
Add to cart
Description
Description
The Human Neutrophil Elastase/ELA2 ELISA Kit (Colorimetric) measures human Neutrophil Elastase/ELA2 in plasma.
Specifications
Specifications
0.16 - 10.0ng/mL
1991
ELISA Kit
1.98pg/mL
Human (Hu)
Neutrophil Elastase/ELA2
ELANE
ELISA
Aluminium pouch(es) with a Microwell Plate coated with polyclonal antibody to human PMN Elastase, HRP-Conjugate anti-alpha1-PI polyclonal antibody, ready to use, Human PMN Elastase Standard lyophilized, 10ng/mL upon reconstitution, Control high, lyophilized, Control low, lyophilized, Sample Diluent, Wash Buffer Concentrate (10x), Substrate Solution (tetramethyl-benzidine), Stop Solution (2M hydrochloric acid), Adhesive Films
96 Tests
Cell culture supernatant, serum, plasma
Storage of components varies. See protocol for specific instructions.
Cancer, Neuroscience
Bone marrow serine protease, EC 3.4.21, EC 3.4.21.37, ELA2granulocyte-derived elastase, elastase 2, neutrophil, elastase, neutrophil expressed, Elastase-2, GE, HLEelastase-2, HNE, Human leukocyte elastase, leukocyte elastase, medullasin, NE, neutrophil el