Learn More
Abnova™ Human NFKBIZ Full-length ORF (NP_113607.1, 1 a.a. - 718 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00064332P01S
Description
This gene is a member of the ankyrin-repeat family and is induced by lipopolysaccharide (LPS). The C-terminal portion of the encoded product which contains the ankyrin repeats, shares high sequence similarity with the I kappa B family of proteins. The latter are known to play a role in inflammatory responses to LPS by their interaction with NF-B proteins through ankyrin-repeat domains. Studies in mouse indicate that this gene product is one of the nuclear I kappa B proteins and an activator of IL-6 production. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MIVDKLLDDSRGGEGLRDAAGGCGLMTSPLNLSYFYGASPPAAAPGACDASCSVLGPSAPGSPGSDSSDFSSASSVSSCGAVESRSRGGARAERQPVEPHMGVGRQQRGPFQGVRVKNSVKELLLHIRSHKQKASGQAVDDFKTQGVNIEQFRELKNTVSYSGKRKGPDSLSDGPACKRPALLHSQFLTPPQTPTPGESMEDVHLNEPKQESSADLLQNIINIKNECSPVSLNTVQVSWLNPVVVPQSSPAEQCQDFHGGQVFSPPQKCQPFQVRGSQQMIDQASLYQYSPQNQHVEQQPHYTHKPTLEYSPFPIPPQSPAYEPNLFDGPESQFCPNQSLVSLLGDQRESENIANPMQTSSSVQQQNDAHLHSFSMMPSSACEAMVGHEMASDSSNTSLPFSNMGNPMNTTQLGKSLFQWQVEQEESKLANISQDQFLSKDADGDTFLHIAVAQGRRALSYVLARKMNALHMLDIKEHNGQSAFQVAVAANQHLIVQDLVNIGAQVNTTDCWGRTPLHVCAEKGHSQVLQAIQKGAVGSNQFVDLEATNYDGLTPLHCAVIAHNAVVHELQRNQQPHSPEVQELLLKNKSLVDTIKCLIQMGAAVEAKDRKSGRTALHLAAEEANLELIRLFLELPSCLSFVNAKAYNGNTALHVAASLQYRLTQLDAVRLLMRKGADPSTRNLENEQPVHLVPDGPVGEQIRRILKGKSIQQRAPPYSpecifications
NP_113607.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
104.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ30225/FLJ34463/IKBZ/INAP/MAIL | |
NFKBIZ | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64332 | |
NFKBIZ (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIVDKLLDDSRGGEGLRDAAGGCGLMTSPLNLSYFYGASPPAAAPGACDASCSVLGPSAPGSPGSDSSDFSSASSVSSCGAVESRSRGGARAERQPVEPHMGVGRQQRGPFQGVRVKNSVKELLLHIRSHKQKASGQAVDDFKTQGVNIEQFRELKNTVSYSGKRKGPDSLSDGPACKRPALLHSQFLTPPQTPTPGESMEDVHLNEPKQESSADLLQNIINIKNECSPVSLNTVQVSWLNPVVVPQSSPAEQCQDFHGGQVFSPPQKCQPFQVRGSQQMIDQASLYQYSPQNQHVEQQPHYTHKPTLEYSPFPIPPQSPAYEPNLFDGPESQFCPNQSLVSLLGDQRESENIANPMQTSSSVQQQNDAHLHSFSMMPSSACEAMVGHEMASDSSNTSLPFSNMGNPMNTTQLGKSLFQWQVEQEESKLANISQDQFLSKDADGDTFLHIAVAQGRRALSYVLARKMNALHMLDIKEHNGQSAFQVAVAANQHLIVQDLVNIGAQVNTTDCWGRTPLHVCAEKGHSQVLQAIQKGAVGSNQFVDLEATNYDGLTPLHCAVIAHNAVVHELQRNQQPHSPEVQELLLKNKSLVDTIKCLIQMGAAVEAKDRKSGRTALHLAAEEANLELIRLFLELPSCLSFVNAKAYNGNTALHVAASLQYRLTQLDAVRLLMRKGADPSTRNLENEQPVHLVPDGPVGEQIRRILKGKSIQQRAPPY | |
RUO | |
NFKBIZ | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.