Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Nicotinamide N-Methyltransferase/NNMT Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $526.00
Specifications
Antigen | Nicotinamide N-Methyltransferase/NNMT |
---|---|
Clone | 954208 |
Dilution | Western Blot 2 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB7736SP
![]() |
R&D Systems
MAB7736SP |
25 μg |
Each for $200.50
|
|
|||||
MAB7736100
![]() |
R&D Systems
MAB7736100 |
100 μg |
Each for $526.00
|
|
|||||
Description
Nicotinamide N-Methyltransferase/NNMT Monoclonal specifically detects Nicotinamide N-Methyltransferase/NNMT in Human samples. It is validated for Western Blot.Specifications
Nicotinamide N-Methyltransferase/NNMT | |
Western Blot 2 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
P40261 | |
4837 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
954208 | |
Western Blot | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
EC 2.1.1.1, nicotinamide N-methyltransferase, Nicotinamide NMethyltransferase, NNMT | |
E.coli-derived recombinant human Nicotinamide N-Methyltransferase/NNMT Met1-Leu264 Accession # P40261 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Nicotinamide N-Methyltransferase/NNMT in direct ELISAs and Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title