Learn More
Abnova™ Human NIPBL Full-length ORF (AAH33847.1, 1 a.a. - 175 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00025836P01S
Description
This gene encodes the homolog of the Drosophila melanogaster Nipped-B gene product and fungal Scc2-type sister chromatid cohesion proteins. The Drosophila protein facilitates enhancer-promoter communication of remote enhancers and plays a role in developmental regulation. It is also homologous to a family of chromosomal adherins with broad roles in sister chromatid cohesion, chromosome condensation, and DNA repair. The human protein has a bipartite nuclear targeting sequence and a putative HEAT repeat. Condensins, cohesins and other complexes with chromosome-related functions also contain HEAT repeats. Mutations in this gene result in Cornelia de Lange syndrome, a disorder characterized by dysmorphic facial features, growth delay, limb reduction defects, and mental retardation. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MKCLPENSAPLIEFANVSQGILLLLMLKQHLKNLCGFSDSKIQKYSPSESAKVYDKAINRKTGVHFHPKQTLDFLRSDMANSKITEEVKRSIVKQYLDFKLLMEHLDPDEEEEEGEVSASTNARNKAITSLLGGGSPKNNTAAETEDDESDGEDRGGGTSGVRRRRSQRISQRITSpecifications
AAH33847.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDLS/CDLS1/DKFZp434L1319/FLJ11203/FLJ12597/FLJ13354/FLJ13648/FLJ44854/IDN3/IDN3-B | |
NIPBL | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
25836 | |
NIPBL (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKCLPENSAPLIEFANVSQGILLLLMLKQHLKNLCGFSDSKIQKYSPSESAKVYDKAINRKTGVHFHPKQTLDFLRSDMANSKITEEVKRSIVKQYLDFKLLMEHLDPDEEEEEGEVSASTNARNKAITSLLGGGSPKNNTAAETEDDESDGEDRGGGTSGVRRRRSQRISQRIT | |
RUO | |
NIPBL | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.