Learn More
Novus Biologicals Human NK2R ELISA Kit (Colorimetric)
This kit recognizes Human TACR2 in samples.
Manufacturer: Novus Biologicals NBP280288
Description
No significant cross-reactivity or interference between Human TACR2 and analogues was observed.Specifications
P21452, P21452 | |
ELISA | |
ELISA Kit (Colorimetric) | |
Storage is content dependent | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
1 Kit | |
Serum, plasma, and other biological fluids |
0.313 to 20 ng/mL | |
6865 | |
0.188 ng/mL | |
Human | |
TACR2 | |
NK2R |