Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NLGN3 Full-length ORF (AAH51715.1, 1 a.a. - 828 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00054413P01S

 View more versions of this product

Catalog No. 89-954-252


Add to cart

Description

Description

This gene encodes a member of a family of neuronal cell surface proteins. Members of this family may act as splice site-specific ligands for beta-neurexins and may be involved in the formation and remodeling of central nervous system synapses. Mutations in this gene may be associated with autism and Asperger syndrome. Multiple transcript variants encoding distinct isoforms have been identified for this gene, but their full length sequences have not been determined. [provided by RefSeq]

Sequence: MWLRLGPPSLSLSPKPTVGRSLCLTLWFLSLALRASTQAPAPTVNTHFGKLRGARVPLPSEILGPVDQYLGVPYAAPPIGEKRFLPPEPPPYWSGIRNATHFPPVCPQNIHTAVPEVMLPVWFTANLDIVATYIQEPNEDCLYLNVYVPTEDGSGAKKQGEDLADNDGDEDEDIRDSGAKPVMVYIHGGSYMEGTGNMIDGSILASYGNVIVITLNYRVGVLGFLSTGDQAAKGNYGLLDQIQALRWVSENIAFFGGDPRRITVFGSGIGASCVSLLTLSHHSEGLFQRAIIQSGSALSSWAVNYQPVKYTSLLADKVGCNVLDTVDMVDCLRQKSAKELVEQDIQPARYHVAFGPVIDGDVIPDDPEILMEQGEFLNYDIMLGVNQGEGLKFVEGVVDPEDGVSGTDFDYSVSNFVDNLYGYPEGKDTLRETIKFMYTDWADRDNPETRRKTLVALFTDHQWVEPSVVTADLHARYGSPTYFYAFYHHCQSLMKPAWSDAAHGDEVPYVFGVPMVGPTDLFPCNFSKNDVMLSAVVMTYWTNFAKTGDPNKPVPQDTKFIHTKANRFEEVAWSKYNPRDQLYLHIGLKPRVRDHYRATKVAFWKHLVPHLYNLHDMFHYTSTTTKVPPPDTTHSSHITRRPNGKTWSTKRPAISPAYSNENAQGSWNGDQDAGPLLVENPRDYSTELSVTIAVGASILFLNVLAFAALYYRKDKRRQEPLRQPSPQRGAWAPELGAAPEEELAALQLGPTHHECEASPPHDTLRLTALPDYTLTLRRSPDDIPLMTPNTITMIPNSLVGLQTLHPYNTFAAGFNSTGLPHSHSTTRV
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
118.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASPGX1/AUTSX1/HNL3/KIAA1480
NLGN3
Yes
AAH51715.1
Liquid
54413
NLGN3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MWLRLGPPSLSLSPKPTVGRSLCLTLWFLSLALRASTQAPAPTVNTHFGKLRGARVPLPSEILGPVDQYLGVPYAAPPIGEKRFLPPEPPPYWSGIRNATHFPPVCPQNIHTAVPEVMLPVWFTANLDIVATYIQEPNEDCLYLNVYVPTEDGSGAKKQGEDLADNDGDEDEDIRDSGAKPVMVYIHGGSYMEGTGNMIDGSILASYGNVIVITLNYRVGVLGFLSTGDQAAKGNYGLLDQIQALRWVSENIAFFGGDPRRITVFGSGIGASCVSLLTLSHHSEGLFQRAIIQSGSALSSWAVNYQPVKYTSLLADKVGCNVLDTVDMVDCLRQKSAKELVEQDIQPARYHVAFGPVIDGDVIPDDPEILMEQGEFLNYDIMLGVNQGEGLKFVEGVVDPEDGVSGTDFDYSVSNFVDNLYGYPEGKDTLRETIKFMYTDWADRDNPETRRKTLVALFTDHQWVEPSVVTADLHARYGSPTYFYAFYHHCQSLMKPAWSDAAHGDEVPYVFGVPMVGPTDLFPCNFSKNDVMLSAVVMTYWTNFAKTGDPNKPVPQDTKFIHTKANRFEEVAWSKYNPRDQLYLHIGLKPRVRDHYRATKVAFWKHLVPHLYNLHDMFHYTSTTTKVPPPDTTHSSHITRRPNGKTWSTKRPAISPAYSNENAQGSWNGDQDAGPLLVENPRDYSTELSVTIAVGASILFLNVLAFAALYYRKDKRRQEPLRQPSPQRGAWAPELGAAPEEELAALQLGPTHHECEASPPHDTLRLTALPDYTLTLRRSPDDIPLMTPNTITMIPNSLVGLQTLHPYNTFAAGFNSTGLPHSHSTTRV
RUO
NLGN3
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit