Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NMRAL1 Full-length ORF (NP_065728.1, 1 a.a. - 299 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$499.00 - $769.00

Specifications

Accession Number NP_065728.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 57407
Molecular Weight (g/mol) 59.7kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-954-319
View Documents
Abnova™
H00057407P01S
10 ug
Each for $499.00
Add to Cart
 
89-954-320
View Documents
Abnova™
H00057407P01L
25 ug
Each for $769.00
Add to Cart
 
Description

Description

Sequence: MVDKKLVVVFGGTGAQGGSVARTLLEDGTFKVRVVTRNPRKKAAKELRLQGAEVVQGDQDDQVIMELALNGAYATFIVTNYWESCSQEQEVKQGKLLADLARRLGLHYVVYSGLENIKKLTAGRLAAAHFDGKGEVEEYFRDIGVPMTSVRLPCYFENLLSHFLPQKAPDGKSYLLSLPTGDVPMDGMSVSDLGPVVLSLLKMPEKYVGQNIGLSTCRHTAEEYAALLTKHTRKVVHDAKMTPEDYEKLGFPGARDLANMFRFYALRPDRDIELTLRLNPKALTLDQWLEQHKGDFNLL
Specifications

Specifications

NP_065728.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
59.7kDa
Glutathione Sepharose 4 Fast Flow
MVDKKLVVVFGGTGAQGGSVARTLLEDGTFKVRVVTRNPRKKAAKELRLQGAEVVQGDQDDQVIMELALNGAYATFIVTNYWESCSQEQEVKQGKLLADLARRLGLHYVVYSGLENIKKLTAGRLAAAHFDGKGEVEEYFRDIGVPMTSVRLPCYFENLLSHFLPQKAPDGKSYLLSLPTGDVPMDGMSVSDLGPVVLSLLKMPEKYVGQNIGLSTCRHTAEEYAALLTKHTRKVVHDAKMTPEDYEKLGFPGARDLANMFRFYALRPDRDIELTLRLNPKALTLDQWLEQHKGDFNLL
RUO
NMRAL1
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
57407
NMRAL1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ25918/HSCARG/SDR48A1
NMRAL1
Recombinant
wheat germ expression system
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit