Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human NRG1 Isoform GGF2 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Neuregulin-1 Isoform GGF2 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Neuregulin-1 Isoform GGF2 Polyclonal specifically detects Neuregulin-1 Isoform GGF2 in Human samples. It is validated for Western Blot.Specifications
Neuregulin-1 Isoform GGF2 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
GGF2, ARIA, GGF, GGF2, Heregulin-1, HGL, HRG, HRG1, HRG1-alpha, HRG1-beta 1, HRGA, MST131, MSTP131, NDF, Neuregulin1, Neuregulin1 Isoform GGF2, NRG1, NRG1-IT2, SMDF | |
NRG1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
NP_039256 | |
3084 | |
E. coli-derived recombinant human NRG1 Isoform GGF2 Gly51-Cys248 Accession # NP_039256 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human NRG1 Isoform GGF2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) NRG-1 alpha EGF domain, rhNRG1-beta 1 extracellular domain, rhNRG1-beta 1 EGF domain, rhSMDF, and recombinant mouse NRG3 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title