Learn More
Abnova™ Human NSFL1C Full-length ORF (NP_057227.2, 1 a.a. - 370 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_057227.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 55968 |
Molecular Weight (g/mol) | 67kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-954-676
|
Abnova™
H00055968P01L |
25 ug |
Each for $769.00
|
|
|||||
89-954-675
|
Abnova™
H00055968P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
N-ethylmaleimide-sensitive factor (NSF) and valosin-containing protein (p97) are two ATPases known to be involved in transport vesicle/target membrane fusion and fusions between membrane compartments. A trimer of the protein encoded by this gene binds a hexamer of cytosolic p97 and is required for p97-mediated regrowth of Golgi cisternae from mitotic Golgi fragments. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MAAERQEALREFVAVTGAEEDRARFFLESAGWDLQIALASFYEDGGDEDIVTISQATPSSVSRGTAPSDNRVTSFRDLIHDQDEDEEEEEGQRFYAGGSERSGQQIVGPPRKKSPNELVDDLFKGAKEHGAVAVERVTKSPGETSKPRPFAGGGYRLGAAPEEESAYVAGEKRQHSSQDVHVVLKLWKSGFSLDNGELRSYQDPSNAQFLESIRRGEVPAELRRLAHGGQVNLDMEDHRDEDFVKPKGAFKAFTGEGQKLGSTAPQVLSTSSPAQQAENEAKASSSILIDESEPTTNIQIRLADGGRLVQKFNHSHRISDIRLFIVDARPAMAATSFILMTTFPNKELADESQTLKEANLLNAVIVQRLTSpecifications
NP_057227.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
67kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAERQEALREFVAVTGAEEDRARFFLESAGWDLQIALASFYEDGGDEDIVTISQATPSSVSRGTAPSDNRVTSFRDLIHDQDEDEEEEEGQRFYAGGSERSGQQIVGPPRKKSPNELVDDLFKGAKEHGAVAVERVTKSPGETSKPRPFAGGGYRLGAAPEEESAYVAGEKRQHSSQDVHVVLKLWKSGFSLDNGELRSYQDPSNAQFLESIRRGEVPAELRRLAHGGQVNLDMEDHRDEDFVKPKGAFKAFTGEGQKLGSTAPQVLSTSSPAQQAENEAKASSSILIDESEPTTNIQIRLADGGRLVQKFNHSHRISDIRLFIVDARPAMAATSFILMTTFPNKELADESQTLKEANLLNAVIVQRLT | |
RUO | |
NSFL1C | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
55968 | |
NSFL1C (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC3347/UBX1/UBXD10/UBXN2C/dJ776F14.1/p47 | |
NSFL1C | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.