Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human NUDT14 Partial ORF (NP_803877.2, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00256281Q01L
Description
UDP-glucose (UDPG) acts as the sugar donor in numerous glycosylation reactions, including those involved in the production of glycogen. NUDT14 is a UDPG pyrophosphatase (EC 3.6.1.45) that hydrolyzes UDPG to produce glucose 1-phosphate and UMP (Yagi et al., 2003 [PubMed 12429023]).[supplied by OMIM]
Sequence: MERIEGASVGRCAASPYLRPLTLHYRQNGAQKSWDFMKTHDSVTVLLFNSSRRSLVLVKQFRPAVYAGEVERRFPGSLAAVDQDGPRELQPALPGSAGVTSpecifications
NP_803877.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MERIEGASVGRCAASPYLRPLTLHYRQNGAQKSWDFMKTHDSVTVLLFNSSRRSLVLVKQFRPAVYAGEVERRFPGSLAAVDQDGPRELQPALPGSAGVT | |
RUO | |
NUDT14 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
256281 | |
NUDT14 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
UGPP/UGPPase | |
NUDT14 | |
Recombinant | |
wheat germ expression system |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction