Learn More
Abnova™ Human NXF5 Partial ORF (NP_116564, 271 a.a. - 370 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00055998Q01L
Description
This gene is one member of a family of nuclear RNA export factor genes. The encoded protein can bind RNA, and is implicated in mRNA nuclear export. However, this protein has lost several C-terminal protein domains found in other family members that are required for export activity, and may be an evolving pseudogene. Alternatively spliced transcript variants have been described, but most are candidates for nonsense-mediated decay (NMD) and may not express proteins in vivo. [provided by RefSeq]
Sequence: FTGSETLKHLVLQFLQQSNLCKYFKDSRNIKILKDPYLQRKLLKHTKCPRNVDSLSALPETQHDFTSILVDMWYQTVNTCFLPRAGPESQRWWCLLSLKWSpecifications
NP_116564 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
FTGSETLKHLVLQFLQQSNLCKYFKDSRNIKILKDPYLQRKLLKHTKCPRNVDSLSALPETQHDFTSILVDMWYQTVNTCFLPRAGPESQRWWCLLSLKW | |
RUO | |
NXF5 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
55998 | |
NXF5 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NXF5 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.