Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human NXF5 Partial ORF (NP_116564, 271 a.a. - 370 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name NXF5 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 36.74kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-954-886
View Documents
Abnova™
H00055998Q01L
25 ug
Each for $709.50
Add to cart
 
89-954-885
View Documents
Abnova™
H00055998Q01S
10 ug
Each for $460.50
Add to cart
 
Description

Description

This gene is one member of a family of nuclear RNA export factor genes. The encoded protein can bind RNA, and is implicated in mRNA nuclear export. However, this protein has lost several C-terminal protein domains found in other family members that are required for export activity, and may be an evolving pseudogene. Alternatively spliced transcript variants have been described, but most are candidates for nonsense-mediated decay (NMD) and may not express proteins in vivo. [provided by RefSeq]

Sequence: FTGSETLKHLVLQFLQQSNLCKYFKDSRNIKILKDPYLQRKLLKHTKCPRNVDSLSALPETQHDFTSILVDMWYQTVNTCFLPRAGPESQRWWCLLSLKW
Specifications

Specifications

NXF5 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
36.74kDa
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit