Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human OATL1 Partial ORF (NP_002527.1, 302 a.a. - 410 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00004943Q01S

 View more versions of this product

Catalog No. 89-954-920


Add to cart

Description

Description

This gene encodes a protein with a TBC domain and may function as a Rab GTPase activating protein. This gene was previously known as ornithine aminotransferase-like 1, but has no similarity to ornithine aminotransferase. [provided by RefSeq]

Sequence: YAGPEDGPHLRALHDLLTTYAVTHPQVSYCQGMSDLASPILAVMDHEGHAFVCFCGIMKRLAANFHPDGRAMATKFAHLKLLLRHADPDFYQYLQEAGADDLFFCYRWL
Specifications

Specifications

NP_002527.1
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.73kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
YAGPEDGPHLRALHDLLTTYAVTHPQVSYCQGMSDLASPILAVMDHEGHAFVCFCGIMKRLAANFHPDGRAMATKFAHLKLLLRHADPDFYQYLQEAGADDLFFCYRWL
RUO
TBC1D25
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
4943
OATL1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MG81|MGC126866|MGC126868|MGC149731|MGC149732|OATL1
TBC1D25
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit