Learn More
Abnova™ Human OAZ3 Full-length ORF (AAH73949.1, 1 a.a. - 190 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH73949.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 51686 |
Molecular Weight (g/mol) | 48.1kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-954-924
|
Abnova™
H00051686P01S |
10 ug |
Each for $499.00
|
|
|||||
89-954-925
|
Abnova™
H00051686P01L |
25 ug |
Each for $769.00
|
|
|||||
Description
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis. The ornithine decarboxylase antizymes play a role in the regulation of polyamine synthesis by binding to and inhibiting ornithine decarboxylase. Antizyme expression is auto-regulated by polyamine-enhanced translational frameshifting. In contrast to antizymes 1 and 2, which are widely expressed throughout the body, the expression of this gene product (antizyme 3) is restricted to testis germ cells, and thus it is a possible candidate for heritable forms of human male infertility. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MTVPWRPGKRRITYKEEEDLTLQPRPASSAPESLVGLQEGKSTEQGNHDQLKELYSAGNLTVLATDPLLHQDPVQLDFHFRLTSQTSAHWHGLLCDRRLFLDIPYQALDQGNRESLTATLEYVEEKTNVDSVFVNFQNDRNDRGALLRAFSYMGFEVVRPDHPALPPLDNVIFMVYPLERDVGHLPSEPPSpecifications
AAH73949.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
48.1kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTVPWRPGKRRITYKEEEDLTLQPRPASSAPESLVGLQEGKSTEQGNHDQLKELYSAGNLTVLATDPLLHQDPVQLDFHFRLTSQTSAHWHGLLCDRRLFLDIPYQALDQGNRESLTATLEYVEEKTNVDSVFVNFQNDRNDRGALLRAFSYMGFEVVRPDHPALPPLDNVIFMVYPLERDVGHLPSEPP | |
RUO | |
OAZ3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
51686 | |
OAZ3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AZ3/OAZ-t/TISP15 | |
OAZ3 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.