Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Oncostatin M (HEK-293-expressed) Recombinant Protein

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

$219.00 - $659.00

Specifications

Gene Alias MGC20461, oncostatin M, oncostatin-M
Source Human embryonic kidney cell,HEK293-derived human Oncostatin M/OSM protein Ala26-Arg221
Conjugate Unconjugated
Recombinant Yes
Gene ID (Entrez) 5008
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
84-75O-M010
View Documents
R&D Systems™
8475OM010
10μg
Each for $219.00
Add to cart
 
84-75O-M050
View Documents
R&D Systems™
8475OM050
50μg
Each for $659.00
Add to cart
 
Description

Description

The Recombinant Human Oncostatin M (HEK-293-expressed) Protein is derived from HEK293. The Recombinant Human Oncostatin M (HEK-293-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications

Specifications

MGC20461, oncostatin M, oncostatin-M
Unconjugated
5008
Human
OSM
<0.10 EU per 1μg of the protein by the LAL method.
Bioactivity
Human embryonic kidney cell,HEK293-derived human Oncostatin M/OSM protein Ala26-Arg221
Yes
>95%, by SDS-PAGE with Silver Staining
Observed M.W.: 27-35kDa, reducing conditions, Theoretical M.W.: 22kDa
Measured in a cell proliferation assay using TF-1 human erythroleukemic cells. Kitamura, T. et al. (1989) J. Cell Physiol. 140:323. The ED50 for this effect is typically 0.05-0.3ng/mL.
P13725
Human embryonic kidney cell, HEK293-derived Ala26-Arg221
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit