Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ORC3L Full-length ORF (NP_862820.1, 1 a.a. - 712 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023595P01S

 View more versions of this product

Catalog No. 89-955-295


Add to cart

Description

Description

The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. Studies of a similar gene in Drosophila suggested a possible role of this protein in neuronal proliferation and olfactory memory. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene. [provided by RefSeq]

Sequence: MATSSMSKGCFVFKPNSKKRKISLPIEDYFNKGKNEPEDSKLRFETYQLIWQQMKSENERLQEELNKNLFDNLIEFLQKSHSGFQKNSRDLGGQIKLREIPTAALVLGVNVTDHDLTFGSLTEALQNNVTPYVVSLQAKDCPDMKHFLQKLISQLMDCCVDIKSKEEESVHVTQRKTHYSMDSLSSWYMTVTQKTDPKMLSKKRTTSSQWQSPPVVVILKDMESFATKVLQDFIIISSQHLHEFPLILIFGIATSPIIIHRLLPHAVSSLLCIELFQSLSCKEHLTTVLDKLLLTTQFPFKINEKVLQVLTNIFLYHDFSVQNFIKGLQLSLLEHFYSQPLSVLCCNLPEAKRRINFLSNNQCENIRRLPSFRRYVEKQASEKQVALLTNERYLKEETQLLLENLHVYHMNYFLVLRCLHKFTSSLPKYPLGRQIRELYCTCLEKNIWDSEEYASVLQLLRMLAKDELMTILEKCFKVFKSYCENHLGSTAKRIEEFLAQFQSLDAETKEEEDASGSQPKGLQKTDLYHLQKSLLEMKELRRSKKQTKFEVLRENVVNFIDCLVREYLLPPETQPLHEVVYFSAAHALREHLNAAPRIALHTALNNPYYYLKNEALKSEEGCIPNIAPDICIAYKLHLECSRLINLVDWSEAFATVVTAAEKMDANSATSEEMNEIIHARFIRAVSELELLGFIKPTKQKTDHVARLTWGGC
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
108.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
LAT/LATHEO/ORC3
ORC3L
Yes
NP_862820.1
Liquid
23595
ORC3L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATSSMSKGCFVFKPNSKKRKISLPIEDYFNKGKNEPEDSKLRFETYQLIWQQMKSENERLQEELNKNLFDNLIEFLQKSHSGFQKNSRDLGGQIKLREIPTAALVLGVNVTDHDLTFGSLTEALQNNVTPYVVSLQAKDCPDMKHFLQKLISQLMDCCVDIKSKEEESVHVTQRKTHYSMDSLSSWYMTVTQKTDPKMLSKKRTTSSQWQSPPVVVILKDMESFATKVLQDFIIISSQHLHEFPLILIFGIATSPIIIHRLLPHAVSSLLCIELFQSLSCKEHLTTVLDKLLLTTQFPFKINEKVLQVLTNIFLYHDFSVQNFIKGLQLSLLEHFYSQPLSVLCCNLPEAKRRINFLSNNQCENIRRLPSFRRYVEKQASEKQVALLTNERYLKEETQLLLENLHVYHMNYFLVLRCLHKFTSSLPKYPLGRQIRELYCTCLEKNIWDSEEYASVLQLLRMLAKDELMTILEKCFKVFKSYCENHLGSTAKRIEEFLAQFQSLDAETKEEEDASGSQPKGLQKTDLYHLQKSLLEMKELRRSKKQTKFEVLRENVVNFIDCLVREYLLPPETQPLHEVVYFSAAHALREHLNAAPRIALHTALNNPYYYLKNEALKSEEGCIPNIAPDICIAYKLHLECSRLINLVDWSEAFATVVTAAEKMDANSATSEEMNEIIHARFIRAVSELELLGFIKPTKQKTDHVARLTWGGC
RUO
ORC3L
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit