Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human OSBPL3 Full-length ORF (NP_663162.1, 1 a.a. - 820 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00026031P01L

 View more versions of this product

Catalog No. 89-955-336


Add to cart

Description

Description

This gene encodes a member of the oxysterol-binding protein (OSBP) family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain. Several transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MMSDEKNLGVSQKLVSPSRSTSSCSSKQGSRQDSWEVVEGLRGEMNYTQEPPVQKGFLLKKRKWPLKGWHKRFFYLDKGILKYAKSQTDIEREKLHGCIDVGLSVMSVKKSSKCIDLDTEEHIYHLKVKSEEVFDEWVSKLRHHRMYRQNEIAMFPHEVNHFFSGSTITDSSSGVFDSISSRKRSSISKQNLFQTGSNVSFSCGGETRVPLWLQSSEDMEKCSKDLAHCHAYLVEMSQLLQSMDVLHRTYSAPAINAIQVPKPFSGPVRLHSSNPNLSTLDFGEEKNYSDGSETSSEFSKMQEDLCHIAHKVYFTLRSAFNIMSAEREKLKQLMEQDASSSPSAQVIGLKNALSSENSRDENRALVHQLSNESRLSITDSLSEFFDAQEVLLSPSSSENEISDDDSYVSDISDNLSLDNLSNDLDNERQTLGPVLDSGREAKSRRRTCLPAPCPSSSNISLWNILRNNIGKDLSKVAMPVELNEPLNTLQRLCEELEYSELLDKAAQIPSPLERMVYVAAFAISAYASSYYRAGSKPFNPVLGETYECIREDKGFQFFSEQVSHHPPISACHAESRNFVFWQDVRWKNKFWGKSMEIVPIGTTHVTLPVFGDHFEWNKVTSCIHNILSGQRWIEHYGEIVIKNLHDDSCYCKVNFIKAKYWSTNAHEIEGTVFDRSGKAVHRLFGKWHESIYCGGGSSSACVWRANPMPKGYEQYYSFTQFALELNEMDPSSKSLLPPTDTRFRPDQRFLEEGNLEEAEIQKQRIEQLQRERRRVLEENHVEHQPRFFRKSDDDSWVSNGTYLELRKDLGFSKLDHPVLW
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
120.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp667P1518/KIAA0704/MGC21526/ORP-3/ORP3/OSBP3
OSBPL3
Yes
NP_663162.1
Liquid
26031
OSBPL3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMSDEKNLGVSQKLVSPSRSTSSCSSKQGSRQDSWEVVEGLRGEMNYTQEPPVQKGFLLKKRKWPLKGWHKRFFYLDKGILKYAKSQTDIEREKLHGCIDVGLSVMSVKKSSKCIDLDTEEHIYHLKVKSEEVFDEWVSKLRHHRMYRQNEIAMFPHEVNHFFSGSTITDSSSGVFDSISSRKRSSISKQNLFQTGSNVSFSCGGETRVPLWLQSSEDMEKCSKDLAHCHAYLVEMSQLLQSMDVLHRTYSAPAINAIQVPKPFSGPVRLHSSNPNLSTLDFGEEKNYSDGSETSSEFSKMQEDLCHIAHKVYFTLRSAFNIMSAEREKLKQLMEQDASSSPSAQVIGLKNALSSENSRDENRALVHQLSNESRLSITDSLSEFFDAQEVLLSPSSSENEISDDDSYVSDISDNLSLDNLSNDLDNERQTLGPVLDSGREAKSRRRTCLPAPCPSSSNISLWNILRNNIGKDLSKVAMPVELNEPLNTLQRLCEELEYSELLDKAAQIPSPLERMVYVAAFAISAYASSYYRAGSKPFNPVLGETYECIREDKGFQFFSEQVSHHPPISACHAESRNFVFWQDVRWKNKFWGKSMEIVPIGTTHVTLPVFGDHFEWNKVTSCIHNILSGQRWIEHYGEIVIKNLHDDSCYCKVNFIKAKYWSTNAHEIEGTVFDRSGKAVHRLFGKWHESIYCGGGSSSACVWRANPMPKGYEQYYSFTQFALELNEMDPSSKSLLPPTDTRFRPDQRFLEEGNLEEAEIQKQRIEQLQRERRRVLEENHVEHQPRFFRKSDDDSWVSNGTYLELRKDLGFSKLDHPVLW
RUO
OSBPL3
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit