Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Osteoadherin/OSAD Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | Osteoadherin/OSAD/OMD |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Osteoadherin/OSAD/OMD Polyclonal specifically detects Osteoadherin/OSAD/OMD in Human samples. It is validated for Western Blot.Specifications
Osteoadherin/OSAD/OMD | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
Keratan sulfate proteoglycan osteomodulin, KSPG osteomodulin, OMD, OSAD, Osteoadherin, osteoadherin proteoglycan, osteomodulin, SLRR2C, SLRR2Costeoadherin | |
OMD | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q99983.1 | |
4958 | |
Mouse myeloma cell line NS0-derived recombinant human Osteoadherin/OSAD Gln21-Glu421 (Phe67Leu) Accession # Q99983.1 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Osteoadherin/OSAD in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant mouse Osteoadherin is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title