Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Osteopontin/OPN Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2020.00
Specifications
Antigen | Osteopontin/OPN |
---|---|
Clone | 223112 |
Concentration | LYOPH |
Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
Applications | ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB14332SP
![]() |
R&D Systems
MAB14332SP |
25 μg |
Each for $200.50
|
|
|||||
MAB14332100
![]() |
R&D Systems
MAB14332100 |
100 μg |
Each for $549.50
|
|
|||||
MAB14332500
![]() |
R&D Systems
MAB14332500 |
500 μg |
Each for $2,020.00
|
|
|||||
Description
Osteopontin/OPN Monoclonal specifically detects Osteopontin/OPN in Human samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
Osteopontin/OPN | |
LYOPH | |
ELISA | |
Unconjugated | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BNSP, Bone sialoprotein 1, Eta-1, MGC110940, Nephropontin, OPN, osteopontin, secreted phosphoprotein 1bone sialoprotein I, early T-lymphocyteactivation 1), secreted phosphoprotein-1 (osteopontin, bone sialoprotein), Spp1, SPP-1, SPP1/CALPHA1 fusion, Urinary stone protein, uropontin | |
SPP1 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
223112 | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Monoclonal | |
Mouse | |
Human | |
NP_000573.1 | |
6696 | |
Mouse myeloma cell line NS0-derived recombinant human Osteopontin Ile17-Asn300 Accession # NP_000573.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Osteopontin in ELISAs. In ELISAs, this antibody does not cross-react with recombinant mouse Osteopontin. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title