Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Osteoprotegerin/TNFRSF11B Alexa Fluor™ 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB8052V100UG
This item is not returnable.
View return policy
Description
Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for Flow Cytometry.Specifications
Osteoprotegerin/TNFRSF11B | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
OCIF, OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TNFRSF11B, TR1, tumor necrosis factor receptor superfamily, member 11b | |
Mouse myeloma cell line, NS0-derived human Osteoprotegerin/TNFRSF11B, Glu22-Leu401, Accession # AAB53709.1 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
155321 | |
Flow Cytometry 0.25-1 ug/106 cells | |
AAB53709.1 | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
4982 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied. | |
IgG1 |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction