Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human OTUB2 Full-length ORF (NP_075601.1, 1 a.a. - 234 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00078990P01S

 View more versions of this product

Catalog No. 89-955-400


Add to cart

Description

Description

Otubains are deubiquitylating cysteine proteases (DUBs; see MIM 602519) that belong to the ovarian tumor (OTU) protein superfamily. Like other DUBs, otubains cleave proteins precisely at the ubiquitin (UB; see MIM 191339)-protein bond.[supplied by OMIM]

Sequence: MSETSFNLISEKCDILSILRDHPENRIYRRKIEELSKRFTAIRKTKGDGNCFYRALGYSYLESLLGKSREIFKFKERVLQTPNDLLAAGFEEHKFRNFFNAFYSVVELVEKDGSVSSLLKVFNDQSASDHIVQFLRLLTSAFIRNRADFFRHFIDEEMDIKDFCTHEVEPMATECDHIQITALSQALSIALQVEYVDEMDTALNHHVFPEAATPSVYLLYKTSHYNILYAADKH
Specifications

Specifications

NP_075601.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
53.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C14orf137|FLJ21916|MGC3102|OTB2|OTU2
OTUB2
Yes
wheat germ expression system
Liquid
78990
OTUB2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSETSFNLISEKCDILSILRDHPENRIYRRKIEELSKRFTAIRKTKGDGNCFYRALGYSYLESLLGKSREIFKFKERVLQTPNDLLAAGFEEHKFRNFFNAFYSVVELVEKDGSVSSLLKVFNDQSASDHIVQFLRLLTSAFIRNRADFFRHFIDEEMDIKDFCTHEVEPMATECDHIQITALSQALSIALQVEYVDEMDTALNHHVFPEAATPSVYLLYKTSHYNILYAADKH
RUO
OTUB2
Wheat Germ (in vitro)
GST
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit