Learn More
Cell Biolabs IncSupplier Diversity Partner HUMAN OXLDL ELISA MDA-LDL 96W  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA369
This OxiSelect Human Oxidized LDL ELISA Kit selectively measures MDA-LDL in plasma, serum or other biological fluid samples. Includes a copper oxidized LDL standard. 96 wells including standards and unknowns.
Catalog No. 50-673-567