Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human p15INK4b/CDKN2B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6798SP
This item is not returnable.
View return policy
Description
p15INK4b/CDKN2B Monoclonal specifically detects p15INK4b/CDKN2B in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
p15INK4b/CDKN2B | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 3-25 ug/mL | |
P42772 | |
CDKN2B | |
E. coli-derived recombinant human p15INK4b/CDKN2B Arg2-Asp138 Accession # P42772 | |
25 μg | |
Primary | |
Detects human p15INK4b/CDKN2B in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant human (rh) p16INK4a and no cross-reactivity with rhp18INK4c or rhp19INK4d is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
651308 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CDK4I, CDKN2B, cyclin-dependent kinase inhibitor 2B (p15, inhibits CDK4), cyclin-dependent kinases 4 and 6 binding protein, INK4BCDK4B inhibitor, MTS2, MTS-2, MTS2CDK inhibitory protein, Multiple tumor suppressor 2, p14_CDK inhibitor, p14_INK4B, p14-INK4b, p15 CDK inhibitor, p15_INK4B, P15cyclin-dependent kinase 4 inhibitor B, p15INK4b, p15-INK4b, TP15 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1030 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction