Learn More
Novus Biologicals™ Human p27/Kip1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP266683
Catalog No. NBP266683
Add to cart
Description
Description
The Human p27/Kip1 ELISA Kit (Colorimetric) measures human p27/Kip1 in serum, plasma and other biological fluids.
Specifications
Specifications
15.63 - 1000 pg/mL
This kit recognizes Human P27 in samples. No significant cross-reactivity or interference between human P27 and analogues was observed.
1027
Human
1 Kit
Storage is content dependent.
9.38 pg/mL
ELISA
CDKN1B
ELISA Kit (Colorimetric)
Biological Fluids,Plasma,Serum
p27/Kip1