Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human P4HTM Full-length ORF (NP_808807.2, 1 a.a. - 563 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00054681P01S

 View more versions of this product

Catalog No. 89-955-511


Add to cart

Description

Description

The product of this gene belongs to the family of prolyl 4-hydroxylases. This protein is a prolyl hydroxylase that may be involved in the degradation of hypoxia-inducible transcription factors under normoxia. It plays a role in adaptation to hypoxia and may be related to cellular oxygen sensing. Alternatively spliced variants encoding different isoforms have been identified. [provided by RefSeq]

Sequence: MAAAAVTGQRPETAAAEEASRPQWAPPDHCQAQAAAGLGDGEDAPVRPLCKPRGICSRAYFLVLMVFVHLYLGNVLALLLFVHYSNGDESSDPGPQHRAQGPGPEPTLGPLTRLEGIKVGHERKVQLVTDRDHFIRTLSLKPLLFEIPGFLTDEECRLIIHLAQMKGLQRSQILPTEEYEEAMSTMQVSQLDLFRLLDQNRDGHLQLREVLAQTRLGNGWWMTPESIQEMYAAIKADPDGDGVLSLQEFSNMDLRDFHKYMRSHKAESSELVRNSHHTWLYQGEGAHHIMRAIRQRVLRLTRLSPEIVELSEPLQVVRYGEGGHYHAHVDSGPVYPETICSHTKLVANESVPFETSCRQVSPNWGLPSILRPGTPMTQAQPCTVGVPLGMGPGDHWVIPVSPWEHPQLGTCSVPPLPYSYMTVLFYLNNVTGGGETVFPVADNRTYDEMSLIQDDVDLRDTRRHCDKGNLRVKPQQGTAVFWYNYLPDGQGWVGDVDDYSLHGGCLVTRGTKWIANNWINVDPSRARQALFQQEMARLAREGGTDSQPEWALDRAYRDARVEL
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
89.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EGLN4/HIFPH4/P4H-TM/PH-4/PH4/PHD4
P4HTM
Yes
NP_808807.2
Liquid
54681
P4HTM (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAAAVTGQRPETAAAEEASRPQWAPPDHCQAQAAAGLGDGEDAPVRPLCKPRGICSRAYFLVLMVFVHLYLGNVLALLLFVHYSNGDESSDPGPQHRAQGPGPEPTLGPLTRLEGIKVGHERKVQLVTDRDHFIRTLSLKPLLFEIPGFLTDEECRLIIHLAQMKGLQRSQILPTEEYEEAMSTMQVSQLDLFRLLDQNRDGHLQLREVLAQTRLGNGWWMTPESIQEMYAAIKADPDGDGVLSLQEFSNMDLRDFHKYMRSHKAESSELVRNSHHTWLYQGEGAHHIMRAIRQRVLRLTRLSPEIVELSEPLQVVRYGEGGHYHAHVDSGPVYPETICSHTKLVANESVPFETSCRQVSPNWGLPSILRPGTPMTQAQPCTVGVPLGMGPGDHWVIPVSPWEHPQLGTCSVPPLPYSYMTVLFYLNNVTGGGETVFPVADNRTYDEMSLIQDDVDLRDTRRHCDKGNLRVKPQQGTAVFWYNYLPDGQGWVGDVDDYSLHGGCLVTRGTKWIANNWINVDPSRARQALFQQEMARLAREGGTDSQPEWALDRAYRDARVEL
RUO
P4HTM
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit